Learn More
Justrite™ Dip Tanks
Product Code. 11994318
Change view
Click to view available options
Capacity (Metric):
4 L
8 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11994318 4 L Each
11953639 8 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11994318 Supplier Justrite™ Supplier No. 27611
  • ONLINE EXCLUSIVE PRICE
    155.40€
    133.83€ / Pack of 1
    Save 21.57€
    13% Off
Estimated Shipment: 03-12-2026
Specifications
Type Dip Tank
Material Steel
Capacity (Metric) 4 L
Height (Metric) 140 mm
For Use With (Equipment) Cleaning Parts
No. per Pack 1 Each