Learn More
Justrite™ Dip Tanks
Product Code. 11953639
Change view
Click to view available options
Capacity (Metric):
4 L
8 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11953639 8 L Each
11994318 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11953639 Supplier Justrite™ Supplier No. 27612
  • ONLINE EXCLUSIVE PRICE
    161.70€
    137.57€ / Pack of 1
    Save 24.13€
    14% Off
Estimated Shipment: 21-08-2026
Specifications
Type Dip Tank
Material Steel
Capacity (Metric) 8 L
Height (Metric) 254 mm
For Use With (Equipment) Cleaning Parts
No. per Pack 1 Each