Learn More
Thermo Scientific™ Dionex™ Ultra-Low Dead Volume Union
Product Code. 11392133 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Nano ES
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11392133 Nano ES Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11392133 Supplier Thermo Scientific™ Supplier No. 160494
Estimated Shipment: 10-11-2026
Thermo Scientific™ Ultra-Low Dead Volume Union is for nano electrospray ionization.
Thermo Scientific™ Ultra-Low Dead Volume Union is for nano electrospray ionization.
  • Dead volume of <7nL
  • With two nuts and two ferrules
  • I.D.: 1/16in.
TRUSTED_SUSTAINABILITY
Specifications
Description Union
For Use With (Equipment) Nano ES
Diameter (English) Outer 1/16 in.
Product Line nanoLC