Learn More
Afora™ Dimroth Condenser
Provide additional surface area for highly efficient cooling with the Afora™ Dimroth Compressor, which pressurizes and heats refrigerant.
235.00€
Description
- Cold finger
- Coiled
- Fluid-cooled
- Internal double spiral
- Often used with rotary evaporators
Specifications
UNE 400316 | |
Dimroth Condenser |