Learn More
Afora™ Dimroth Condenser
Product Code. 12080088
Change view
Click to view available options
Standard Taper Joint No:
29/32
Weight (Metric) Shipping:
0.01 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Weight (Metric) Shipping Unit Size
12080088 29/32 0.01 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12080088 Supplier Afora™ Supplier No. 1602/E
  • ONLINE EXCLUSIVE PRICE
    270.49€
    91.27€ / Pack of 1
    Save 179.22€
    66% Off
Estimated Shipment: 14-12-2026
Provide additional surface area for highly efficient cooling with the Afora™ Dimroth Compressor, which pressurizes and heats refrigerant.
  • Cold finger
  • Coiled
  • Fluid-cooled
  • Internal double spiral
  • Often used with rotary evaporators
Specifications
Certifications/Compliance UNE 400316
Standard Taper Joint No 29/32
Weight (Metric) Shipping 0.01 kg
Type Dimroth Condenser