Learn More
Afora™ Dimroth Condenser
Product Code. 11925869
Change view
Click to view available options
Standard Taper Joint No:
14/23
19/26
Height (Metric):
293 mm
316 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Height (Metric) unitSize
11925869 19/26 316 mm Each
11925879 14/23 293 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11925869 Supplier Afora™ Supplier No. 1592/E
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides more effective cooling than conventional coil condensers. Afora™ Dimroth Condenser is a cold finger, coiled, fluid-cooled condenser.
  • Internal double spiral
  • Coolant inlet and outlet reside at the top of the condenser
  • Often used with rotary evaporators
  • Complies with UNE 400316
Specifications
Certifications/Compliance UNE 400316
Description Exchange Surface: 250cm2
Height (Metric) 316 mm
Length (Metric) Jacket 200 mm
Standard Taper Joint No 19/26
Type Condenser