missing translation for 'onlineSavingsMsg'
Learn More

Dimethylbenzyl-(3-sulfopropyl)ammonium, Inner Salt, TRC

Product Code. 30375046
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375046 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375046 Supplier Toronto Research Chemicals Supplier No. TRCD4652505G
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 81239-45-4
Molecular Formula C12 H19 N O3 S
Quantity 5 g
InChI Formula InChI=1S/C12H19NO3S/c1-13(2,9-6-10-17(14,15)16)11-12-7-4-3-5-8-12/h3-5,7-8H,6,9-11H2,1-2H3
SMILES C[N+](C)(CCCS(=O)(=O)[O-])Cc1ccccc1
IUPAC Name 3-[benzyl(dimethyl)azaniumyl]propane-1-sulfonate
Molecular Weight (g/mol) 257.35
Formula Weight 257.1086
Chemical Name or Material Dimethylbenzyl-(3-sulfopropyl)ammonium, Inner Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.