Learn More
Dimethyl Cyclopropane-1,1-dicarboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Dimethyl Cyclopropane-1,1-dicarboxylate |
| CAS | 6914-71-2 |
| Molecular Formula | C7H10O4 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C7H10O4/c1-10-5(8)7(3-4-7)6(9)11-2/h3-4H2,1-2H3 |
| SMILES | COC(=O)C1(CC1)C(=O)OC |
| IUPAC Name | dimethyl cyclopropane-1,1-dicarboxylate |
| Molecular Weight (g/mol) | 158.15 |
| Formula Weight | 158.0579 |