Learn More
Dimethyl 4-Hydroxy-1H-pyrazole-3,5-dicarboxylate, TRC
Product Code. 30336406
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336406 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336406 Supplier Toronto Research Chemicals Supplier No. TRCB42992550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dimethyl 4-Hydroxy-1H-pyrazole-3,5-dicarboxylate
CAS 23705-85-3
Molecular Formula C7 H8 N2 O5
Quantity 50 mg
InChI Formula InChI=1S/C7H8N2O5/c1-13-6(11)3-5(10)4(9-8-3)7(12)14-2/h10H,1-2H3,(H,8,9)
SMILES COC(=O)c1n[nH]c(C(=O)OC)c1O
IUPAC Name dimethyl 4-hydroxy-1H-pyrazole-3,5-dicarboxylate
Molecular Weight (g/mol) 200.149
Formula Weight 200.043