missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethicone, TRC
Specifications
Specifications
| Chemical Name or Material | Dimethicone |
| CAS | 9006-65-9 |
| Molecular Formula | C6 H18 O Si2(C2 H6 O Si)n |
| Quantity | 100 g |
| Synonym | Acesil 350, BY 22-050, Belsil DM 1, Belsil DM 1 Plus, Belsil DM 100, Belsil DM 1000, Belsil DM 200, Belsil DM 35, Belsil DM 5, Clearocast 100, DC 100-350CS, DC 1132, DC 1428, DC 1618, DC 1664, DC 200 Fluid, DC 200-100cS, DC 200-10cS, DC 5-2117, DC 5-7137, DMF 5CS, Dimethicone 245, Dimethicone 350, Dimethicone 5CS, Dimethisil DM 20, Dimeticone, Dow Corning 100-350CS, Dow Corning 1132, Dow Corning 1413, Dow Corning 1618, Dow Corning 1664, Dow Corning 200 Fluid 350 c/s, Dow Corning 200 Fluid 5cSt, Dow Corning 200/100 cSt Fluid, Dow Corning 200/10CST, Dow Corning 200/5 cst, Dow Corning 365, Dow Corning 365 Dimethicone NF Emulsion, Dow Corning 5-2117, Dow Corning 5-7137, Dow Corning 5-7139, E 1049, EY 22-067, Gransil 530, HL 88, HL 999, HZ 201, Hedrin, Jeesile 110, KF 96A50CS, KHS 7, KM 910, M 620, Mirasil DM 20, Mirasil DM 300000, Mirasil DM 350, Mirasil DME 2, Mirasil DME 30, Momentive SE 30 Gum, NYDA, NYDA sensitiv, PMX 200, SH 200C10CS-JASU, Silicone Fluid 350, Silkonoel AK 500, TSF 451-1MA, Viscasil 330000, Viscasil 330M, Viscasil 5M, Visosal 330M, Wacker-Belsil DM 1 Plus, Xiameter 300, Xiameter MEM 1664, Xiameter PMX 200 |
| InChI Formula | InChI=1S/C8H24O2Si3/c1-11(2,3)9-13(7,8)10-12(4,5)6/h1-8H3 |
| SMILES | C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C |
| IUPAC Name | α-Trimethylsilyl-ω-methylpoly[oxy(dimethylsilanediyl)] |
| Molecular Weight (g/mol) | 236.53 |
| Formula Weight | 236.1084 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?