missing translation for 'onlineSavingsMsg'
Learn More

Dimethicone, TRC

Product Code. 30386771
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386771 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386771 Supplier Toronto Research Chemicals Supplier No. TRCD4609405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethicone
CAS 9006-65-9
Molecular Formula C6 H18 O Si2(C2 H6 O Si)n
Quantity 5 g
Synonym Acesil 350, BY 22-050, Belsil DM 1, Belsil DM 1 Plus, Belsil DM 100, Belsil DM 1000, Belsil DM 200, Belsil DM 35, Belsil DM 5, Clearocast 100, DC 100-350CS, DC 1132, DC 1428, DC 1618, DC 1664, DC 200 Fluid, DC 200-100cS, DC 200-10cS, DC 5-2117, DC 5-7137, DMF 5CS, Dimethicone 245, Dimethicone 350, Dimethicone 5CS, Dimethisil DM 20, Dimeticone, Dow Corning 100-350CS, Dow Corning 1132, Dow Corning 1413, Dow Corning 1618, Dow Corning 1664, Dow Corning 200 Fluid 350 c/s, Dow Corning 200 Fluid 5cSt, Dow Corning 200/100 cSt Fluid, Dow Corning 200/10CST, Dow Corning 200/5 cst, Dow Corning 365, Dow Corning 365 Dimethicone NF Emulsion, Dow Corning 5-2117, Dow Corning 5-7137, Dow Corning 5-7139, E 1049, EY 22-067, Gransil 530, HL 88, HL 999, HZ 201, Hedrin, Jeesile 110, KF 96A50CS, KHS 7, KM 910, M 620, Mirasil DM 20, Mirasil DM 300000, Mirasil DM 350, Mirasil DME 2, Mirasil DME 30, Momentive SE 30 Gum, NYDA, NYDA sensitiv, PMX 200, SH 200C10CS-JASU, Silicone Fluid 350, Silkonoel AK 500, TSF 451-1MA, Viscasil 330000, Viscasil 330M, Viscasil 5M, Visosal 330M, Wacker-Belsil DM 1 Plus, Xiameter 300, Xiameter MEM 1664, Xiameter PMX 200
InChI Formula InChI=1S/C8H24O2Si3/c1-11(2,3)9-13(7,8)10-12(4,5)6/h1-8H3
SMILES C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C
IUPAC Name α-Trimethylsilyl-ω-methylpoly[oxy(dimethylsilanediyl)]
Molecular Weight (g/mol) 236.53
Formula Weight 236.1084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.