Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V6 Pumps
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for V6 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
117.00€ - 219.00€
Specifications
Description | 60mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger |
---|---|
For Use With (Equipment) | V6 Pump |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15218483
|
Trajan Scientific and Medical™
004696 |
50 μL |
145.00€
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15238473
|
Trajan Scientific and Medical™
003696 |
25 μL |
140.00€
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15238493
|
Trajan Scientific and Medical™
006696 |
250 μL |
117.00€
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15238513
|
Trajan Scientific and Medical™
008796 |
5 mL |
132.00€
Each |
Estimated Shipment: 23-04-2024
Log in to see stock. |
|||||
15248503
|
Trajan Scientific and Medical™
008196 |
1 mL |
117.00€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
15278513
|
Trajan Scientific and Medical™
008996 |
10 mL |
173.00€
Each |
Estimated Shipment: 23-04-2024
Log in to see stock. |
|||||
15288483
|
Trajan Scientific and Medical™
005696 |
100 μL |
117.00€
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15288503
|
Trajan Scientific and Medical™
008596 |
2.5 mL |
128.00€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
15288513
|
Trajan Scientific and Medical™
009496 |
25 mL |
219.00€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
15298493
|
Trajan Scientific and Medical™
007696 |
500 μL |
117.00€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Diluter/Dispenser Syringe for V6 Pumps
Specifications
60mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger | |
V6 Pump |