Learn More
Thermo Scientific™ DiluFlux Silicone Tubing
Product Code. 17932992
Change view
Click to view available options
For Use With (Equipment):
Diluflux Sample Dilution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment)
17932992 Diluflux Sample Dilution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17932992 Supplier Thermo Scientific™ Supplier No. DB4119A
Estimated Shipment: 10-12-2026
Silicone tubing 6.4mm (set of 5)
Compatible with many prepared media formats, the easy-to-clean, stainless steel DiluFlux Pro Dilutors are an ideal fit for any food testing laboratory.
Automated media dispensing and sample dilution
  • Compact and easy-to-use
  • Automated, rapid and accurate dispensing
  • Available with one or two pump heads for multiple media dispensing
  • Compatible with Dry-Bags and other media formats
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) Silicone tubing 6.4 mm (set of 5)
For Use With (Equipment) Diluflux Sample Dilution