Learn More
Dihydro-3,4-diphenyl-2,5-furandione;, TRC
Product Code. 30377304
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377304 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30377304 Supplier Toronto Research Chemicals Supplier No. TRCD494055250MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dihydro-3,4-diphenyl-2,5-furandione
CAS 101278-21-1
Molecular Formula C16 H12 O3
Quantity 250 mg
Synonym 2,3-Diphenysuccinic Anhydride
InChI Formula InChI=1S/C16H12O3/c17-15-13(11-7-3-1-4-8-11)14(16(18)19-15)12-9-5-2-6-10-12/h1-10,13-14H
SMILES O=C1OC(=O)C(C1c2ccccc2)c3ccccc3
IUPAC Name 3,4-diphenyloxolane-2,5-dione
Molecular Weight (g/mol) 252.265
Formula Weight 252.079