Learn More
WTW™ Digital Triple Channel Multi Parameter Sonde Kit for WTW™ IDS sensors
GREENER_CHOICE
Product Code. 13446768
Click to view available options
For Use With (Equipment):
IDS Sensors, SensoLyt 900 -P, TetraCon 925 -P, FDO 925 -P
Unit Size:
Each
Product Code. 13446768
Brand: WTW™ 401206
Only null left
Specifications
For Use With (Application) Designed for depth measurements to 100m and for pH, conductivity, DO measurement
Includes Built-in pressure sensor, guard, weight, protection cap, instruction manual, CD- ROM with software, 1.5m connection cable of AS IDS/1.5, carry case including SensoLyt™ 900 -P, TetraCon™ 925 -P
For Use With (Equipment) IDS Sensors, SensoLyt 900 -P, TetraCon 925 -P, FDO 925 -P