Learn More
Aalborg™ Digital Mass Flowmeter
Product Code. 15359904
Click to view available options
Airflow Rate:
0 to 10 mL/min.
20 to 50 L/min.
Model:
Digital Mass Flowmeter
Stainless Steel Mass Flowmeter
Total Air Mass Flowmeter
Unit Size:
Each
Product Code. 15359904
Brand: Aalborg™ DFMS010581
Only null left
Add to basket
Add to basket
Use a local four button teypad and LCD to access all features of the flow meter. Aalborg™ Digital Mass Flowmeter supports automatic sensor zero offset adjustments.
  • Flow Alarm
    • High and Low gas flow ALARM limits can be preprogrammed via keypad or remotely via digital interface. ALARM conditions become true when the current flow reading is equal or higher/lower than corresponding values of high and low alarm levels. Alarm action can be assigned with preset delay interval (0-3600 seconds) to activate the contact closer (separate for High and Low alarm).
  • Pressure Alarm
    • High and Low gas pressure ALARM limits can be preprogrammed via keypad or remotely via digital interface. Pressure alarm conditions become true when the current pressure reading is equal or higher than corresponding values of high pressure alarm settings or equal or lower than corresponding values of low pressure alarm settings. Alarm action can be assigned to activate the contact closer (separate for High and Low pressure alarm).
  • Temperature Alarm
    • High and Low gas temperature ALARM limits can be preprogrammed via keypad or remotely via digital interface. Temperature alarm conditions become true when the current temperature reading is equal or higher than corresponding values of high temperature alarm settings or equal or lower than corresponding values of low temperature alarm settings. Alarm action can be assigned to activate the contact closer (separate for High and Low temperature alarm).
  • Engineering Units
    • The flow set points, measured gas flow and associated totalizer data are scaled directly in engineering units via the front panel keypad or digital interface.
Certifications
NIST
Specifications
Type Flowmeter
Accuracy ±1%
Airflow Rate 0 to 10 mL/min.
Pressure Rating 5 to 100 psig
Connections 0.63 cm (0.25 in.) Standard Compression Fittings
Model Stainless Steel Mass Flowmeter
Repeatability ±0.15% (Full Scale)
Temperature (Metric) 0°C to 50°C