Learn More
Ludwig Schneider Digital Exact-Temp Min/Max Thermometer, -50+70°C res. 0,1°C, 3 m cable, DAkkS-cal. cert. 2 p.
Brand:  Ludwig Schneider 65809/04/2P
Encompass_Preferred
Digital Exact-Temp Min/Max Thermometer, -50+70°C res. 0,1°C, 3 m cable, DAkkS-cal. cert. 2 p.
Product Code. 16608817
  • 277.36€ / Each
Only null left
Estimated Shipment: 22-01-2026