Description
Accessory for Digital Dry Bath Series
Specifications
Specifications
| For Use With (Equipment) | Gilson Digital Dry Bath Series |
| Holds | 96-well PCR Plates (0.2 mL wells) |
| Product Line | Digital Dry Bath Series Block |
| Size | 1 Block |