Learn More
Mitutoyo™ Digimatic DMX-1 D-Sub9 Serial Interface Connector
Description
- Microcontrolled interface
- Connects a single measurment device, with Digimatic output, to an RS-232C computer interface.
- Easily connects to a computer's RS-232C serial port.
- No external power supply is needed, as the small level of power needed is supplied by the handshake lines RTS and DTR.
Specifications
Specifications
| Product Type | DMX-1 D-SUB9 Serial Interface |
| For Use With (Equipment) | Computer/measurement device |
| Length (Metric) | 62 mm |