Learn More
Cole-Parmer™ Digi-Sense™ Dual-Display 2-Channel Jumbo-Digit Digital Clock/Timer
Shop All Cole-Parmer Products
Click to view available options
Time Range:
24 hr.
Unit Size:
Each
Description
- Jumbo digit clock/timer features a dual LCD display that tilts for easy viewing.
- Two channels can be operated and displayed simultaneously.
- Use both channels to count down, or use one channel to count up and the other to count down.
- Built-in clock displays time of day in 12- or 24-hour formats.
- Timer recalls previous time set.
- A 70 dB alarm sounds at the end of the countdown; shuts off at the touch of a button.
- Timer displays time elapsed since the end of a countdown.
- Place timer on tabletop, or mount on wall using the double-sided taped bracket or screws (both are included).
- Includes wall-mounting bracket and one AAA battery.
Specifications
Specifications
| Time Range | 24 hr. |
| Alarms | 1 min. Continuous Beep |
| Accuracy | ±0.01% |
| Resolution | 1 sec. |
| Display | LCD |
| Description | Dual-Display 2-Channel Jumbo-Digit Digital Clock/Timer |
| No. of Channels | 2 |
| Width (Metric) | 9.5 cm |
| Height (English) Display | 1 in. |
| Height (Metric) | 14 cm |