Learn More
Digi-Sense™ Advanced Precalibrated Thermistor Thermometers
Product Code. 15651774
Click to view available options
Model:
Data logging
Not data logging
Probe Type:
2 input
Single input
Unit Size:
Each
Product Code. 15651774
Brand: Digi-Sense™ 2025093
Includes
Protective rubber armor boot with built-in stand, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal. Data logging models 20250-92, -94, and -96 also include a USB cable and software.
Perfect for the lab to production floor
  • Reliable measurements, guaranteed!
  • NIST-traceable certificate included
  • Heavy duty construction to protect against knocks and jars.
  • Super lager LCD for easy viewing of data
  • Accepts a wide variety of probes
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
Warranty
2 Years
Specifications
Disposable Reusable
Height (English) 1 in.
Height (Metric) 2.6 cm
Includes Soft carrying case, lanyard, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
Length (Metric) 178 mm
Material Plastic
Measurement Time <1 sec.
Model Not data logging
Probe Type Single input
Resolution 0.1°
Width (English) 2.25 in.
Product Type Thermistor Thermometer
Data Storage None
Length (English) 7 in.
Max. Temperature (English) 257°F
Max. Temperature (Metric) 125°C
Min. Temperature (English) -40°F
Min. Temperature (Metric) -40°C
Temperature Range (English) -40°F to +257°F
Temperature Range (Metric) -40°C to +125°C
Calibrated NIST-traceable
For Use With (Equipment) 400 Series Thermistor Probes with 1/4 in. phono plug
Battery Type 9V
Accuracy ±0.9°F (±0.5°C) from 32 to 158°F, (0 to 70°C); ±2°F (1°C) outside range
Display Backlit LCD
Width (Metric) 5.7 cm