missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ MacConkey Agar
Click to view available options
:
MacConkey Agar; Quantity: 10 kg/Ea.; Format: Dehydrated
Description
- MacConkey Agar Base is a solid medium recommended for use in qualitative procedures for the cultivation of gram-negative bacilli.
- Peptones provide nitrogenous nutrients and amino acids
- Sodium chloride maintains osmotic equilibrium
- Crystal violet and bile salts inhibit most gram-positive organisms MacConkey Agar Base is used with added carbohydrate to differentiate enteric gram-negative bacilli based on fermentation reactions.
Specifications
Specifications
| Certifications/Compliance | CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP |
| Form | Dehydrated |
| Media Type | MacConkey Agar |
| Formulation | Pancreatic Digest of Gelatin 17 g/L, Peptones (Meat and Casein) 3 g/L, Lactose 10 g/L, Bile Salts 1.5 g/L, Sodium Chloride 5 g/L, Agar 13.5 g/L, Neutral Red 0.03 g/L, Crystal Violet 1 mg/L |
| For Use With (Application) | Differential Isolation of Enteric Bacilli Based on Lactose Fermentation |
| Packaging Type | Pail |
| Quantity | 10 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction