Learn More
BD Difco™Discardit™ Concentric Luer-slip Two-Piece Syringe
Product Code. 10142534
Change view
Click to view available options
Volume (Metric):
2 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10142534 2 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10142534 Supplier BD Supplier No. 300928
  • ONLINE EXCLUSIVE PRICE
    75.73€
    44.91€ / Pack of 100
    Save 30.82€
    40% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Two piece syringes without needles. BD Difco™Discardit™ Concentric Luer-slip Two-Piece Syringe is disposable and sterile.
  • Sterile
  • Single use
  • Two piece syringes
  • Without needles
  • Please see Microlance™ disposable needles range for disposable needles
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Mark, European Directive 93/42 / EEC ( compliance procedure Annex V and VII) Certificates: No. 95 Q 252156 and No. 252231 Q (references 309661 & 309620 ) ; certified by NSAI (No. 0050)
Material Plastic
Color Clear
Latex Free Yes
Product Type Concentric Luer-slip 2-piece Syringe
Disposable Single-use
Graduations 0.2 mL
Quantity 100/Pk.
Volume (Metric) 2 mL
Connection Type Concentric Luer Slip
Graduated Yes