Learn More
BD Difco™ Dehydrated Culture Media: Tryptose Agar
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
Unit Size:
500g
Description
Cultivation of Brucella and a variety of pathogenic microorganisms
Certifications
BAM, CCAM, COMPF
Specifications
Specifications
| Certifications/Compliance | BAM, CCAM, COMPF |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Tryptose Agar |
| Product Type | Agar |
| Formulation | Tryptose 20g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Agar 15g/L |
| For Use With (Application) | Cultivation of a wide variety of fastidious microorganisms |
| Packaging Type | Bottle |
| Quantity | 500 g |