missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine (Nitric Oxide) Adduct, TRC

Product Code. 30378517
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378517 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378517 Supplier Toronto Research Chemicals Supplier No. TRCD443500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylamine (nitric oxide) Adduct
CAS 372965-00-9
Molecular Formula C4 H11 N . C4 H11 N3 O2
Quantity 100 mg
Synonym Ethanamine, N-ethyl-, compd. with 1,1-diethyl-2-hydroxy-2-nitrosohydrazine (1:1), Hydrazine, 1,1-diethyl-2-hydroxy-2-nitroso-, compd. with N-ethylethanamine (1:1) (9CI), 1,1-Diethyl-2-hydroxy-2-nitrosohydrazine diethylammonium salt, Diethylamine NONOate diethylammonium salt
InChI Formula InChI=1S/C4H11N3O2.C4H11N/c1-3-6(4-2)7(9)5-8;1-3-5-4-2/h9H,3-4H2,1-2H3;5H,3-4H2,1-2H3
SMILES CCNCC.CCN(CC)N(O)N=O
IUPAC Name N-(diethylamino)-N-hydroxynitrous amide;N-ethylethanamine
Molecular Weight (g/mol) 206.29
Formula Weight 206.1743
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.