missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 10754281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
500g
5g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10754281 5 g Glass Bottle 5g
10592551 100 g Plastic bottle 100g
10001720 500 g Plastic bottle 500g
10387850 1 kg Plastic bottle 1kg
13458378 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10754281 Supplier Thermo Scientific Acros Supplier No. 155890050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660-68-4
MDL Number MFCD00012499
InChI Key HDITUCONWLWUJR-UHFFFAOYSA-N
Synonym diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl
PubChem CID 10197650
IUPAC Name N-ethylethanamine;hydrochloride
SMILES CCNCC.Cl

Specifications

Melting Point 227°C to 230°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C4H12ClN
Linear Formula (C2H5)2NH·HCl
Quantity 5 g
Molecular Weight (g/mol) 109.597
Formula Weight 109.6
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Diethylamine hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.