missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetylsuccinate, 99%

Product Code. 10696934 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10696934 500 g Glass bottle 500g
10697252 25 g Glass bottle 25g
10793641 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10696934 Supplier Thermo Scientific Acros Supplier No. 113955000
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1115-30-6 | C10H16O5 | 216.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-30-6
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.23
MDL Number MFCD00009157
InChI Key DVSDDICSXBCMQJ-UHFFFAOYSA-N
Synonym diethyl 2-acetylsuccinate, diethyl acetylsuccinate, ethyl acetylsuccinate, diethyl acetosuccinate, butanedioic acid, acetyl-, diethyl ester, acetylsuccinic acid diethyl ester, diethylacetylsuccinate, 1,4-diethyl 2-acetylbutanedioate, succinic acid, acetyl-, diethyl ester, butanedioic acid, 2-acetyl-, 1,4-diethyl ester
PubChem CID 66197
IUPAC Name diethyl 2-acetylbutanedioate
SMILES CCOC(=O)CC(C(=O)C)C(=O)OCC

Specifications

Melting Point -8°C
Color Yellow
Density 1.0800g/mL
Boiling Point 180°C to 183°C (50.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C2H5O2CCH2CH(COCH3)CO2C2H5
Refractive Index 1.4340 to 1.436
Quantity 500 g
Beilstein 03, 801
Specific Gravity 1.08
Solubility Information Solubility in water: 15g/L (20°C)
Formula Weight 216.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl acetylsuccinate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.