Learn More
Diethyl (3-aminopropyl)phosphonate, 95%, Thermo Scientific Chemicals
Product Code. 15478367
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478367 1 g 1g
15488367 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15478367 Supplier Thermo Scientific Alfa Aesar Supplier No. H51874.03
Request Bulk or Custom Format
Diethyl (3-aminopropyl)phosphonate is used in the preparation of tert-Butyl (2-((diethoxyphosphoryl)propyl)amino)-3,4-dioxocyclobut-1- en-1-yl)carbamate by reacting with tert-butyl (2-ethoxy-3,4-dioxocyclobut-1-en-1-yl)carbamate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl (3-aminopropyl)phosphonate is used in the preparation of tert-Butyl (2-((diethoxyphosphoryl)propyl)amino)-3,4-dioxocyclobut-1- en-1-yl)carbamate by reacting with tert-butyl (2-ethoxy-3,4-dioxocyclobut-1-en-1-yl)carbamate.
Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4402-24-8
Molecular Formula C7H18NO3P
Molecular Weight (g/mol) 195.20
MDL Number MFCD11110596
InChI Key DQORFBNFNLHVIM-UHFFFAOYSA-N
Synonym diethyl 3-aminopropyl phosphonate, diethyl 3-aminopropylphosphonate, 3-aminopropyl phosphonic acid diethyl ester, diethyl aminopropyl phosphonate, 3-aminopropylphosphonic acid diethyl ester, 3-aminopropanephosphonic acid diethyl ester, 3-amino-propyl-phosphonic acid diethyl ester, diethyl 3-aminopropyl phosphonate 250mg, phosphonic acid, 3-aminopropyl-, diethyl ester
PubChem CID 78113
IUPAC Name 3-diethoxyphosphorylpropan-1-amine
SMILES CCOP(=O)(CCCN)OCC
Specifications
Density 1.053
Quantity 1 g
UN Number UN2735
Sensitivity Air sensitive
Formula Weight 195.2
Percent Purity 95%
Chemical Name or Material Diethyl (3-aminopropyl)phosphonate