missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3,4-dichlorophenyl)malonate, 95%, Thermo Scientific™

Product Code. p-7028865
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346769 1 g 1g
11356769 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346769 Supplier Thermo Scientific Alfa Aesar Supplier No. H26277.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28751-26-0
Molecular Formula C13H14Cl2O4
Molecular Weight (g/mol) 305.15
MDL Number MFCD02180874
InChI Key GZSMDWSQPZMQDV-UHFFFAOYSA-N
PubChem CID 6710867
IUPAC Name diethyl 2-(3,4-dichlorophenyl)propanedioate
SMILES CCOC(=O)C(C(=O)OCC)C1=CC(Cl)=C(Cl)C=C1

Specifications

Refractive Index 1.5105
Quantity 1 g
Synonym diethyl 3,4-dichlorophenyl malonate, diethyl 2-3,4-dichlorophenyl malonate, diethyl2-3,4-dichlorophenyl malonate, diethyl 2-3,4-dichlorophenyl propanedioate, 1,3-diethyl 2-3,4-dichlorophenyl propanedioate, maybridge1_008739, acmc-1cb1c, 3,4-dichlorophenyl malonic acid diethyl ester, 2-3,4-dichlorophenyl propanedioic acid diethyl ester
Formula Weight 305.16
Percent Purity 95%
Chemical Name or Material Diethyl (3,4-dichlorophenyl)malonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.