Learn More
Diethyl 2-thienylmethylphosphonate, 95%
Product Code. 15472489
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15472489 5 g 5g
15482489 250 mg 250mg
15462489 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15472489 Supplier Thermo Scientific Alfa Aesar Supplier No. H65232.06
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
CAS: 2026-42-8 | C9H15O3PS | 234.25 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2026-42-8
Molecular Formula C9H15O3PS
Molecular Weight (g/mol) 234.25
MDL Number MFCD00128457
InChI Key GOJBUVQDOFHBLF-UHFFFAOYSA-N
Synonym diethyl thiophen-2-ylmethyl phosphonate, diethyl 2-thienylmethyl phosphonate, thiophen-2-ylmethyl-phosphonic acid diethyl ester, diethyl 2-thienylmethylphosphonate, 2-diethoxyphosphorylmethyl thiophene, phosphonic acid, 2-thienylmethyl-, diethyl ester, diethyl thiophen-2-ylmethylphosphonate, ksc207o5j, 2-diethoxyphosphinylmethyl thiophene
PubChem CID 4185897
IUPAC Name 2-(diethoxyphosphorylmethyl)thiophene
SMILES CCOP(=O)(CC1=CC=CS1)OCC
Specifications
Color Yellow
Refractive Index 1.52
Quantity 5 g
Beilstein 1683411
Formula Weight 234.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl 2-thienylmethylphosphonate