missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-Acetamido-2-[2-(4-octylphenyl)-2-oxo-ethyl]malonate, TRC

Product Code. 30372026
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372026 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372026 Supplier Toronto Research Chemicals Supplier No. TRCD4431401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 2-Acetamido-2-[2-(4-octylphenyl)-2-oxo-ethyl]malonate
CAS 268557-49-9
Molecular Formula C23 H36 O3
Quantity 1 g
Synonym (Acetylamino)[2-(4-octylphenyl)-2-oxoethyl]propanedioic Acid Diethyl Ester
InChI Formula InChI=1S/C23H36O3/c1-5-21(25)26-20-9-8-17-16-7-6-15-12-19(24)14(2)13-23(15,4)18(16)10-11-22(17,20)3/h14-18,20H,5-13H2,1-4H3/t14-,15+,16-,17+,18+,20-,22-,23-/m0/s1
SMILES CCC(=O)O[C@H]1CC[C@@H]2[C@@H]3CC[C@@H]4CC(=O)[C@@H](C)C[C@]4(C)[C@@H]3CC[C@]12C
IUPAC Name [(2S,5R,8R,9R,10S,13S,14R,17S)-2,10,13-trimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 360.53
Formula Weight 360.266
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.