Learn More
Diethyl-2,2,2,2',2',2'-d6-amine HCl, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Diethylamine-2,2,2,2',2',2'-D6 Hydrochloride |
| CAS | 1219805-68-1 |
| Molecular Formula | C4 D6 H5 N . Cl H |
| Quantity | 0.5 g |
| Synonym | Ethan-2,2,2-d3-amine, N-(ethyl-2,2,2-d3)-, hydrochloride (1:1) (ACI), 2,2,2-Trideuterio-N-(2,2,2-trideuterioethyl)ethanamine hydrochloride, Diethyl-d6-amine hydrochloride, Diethyl-2,2,2,2',2',2'-d6-amine hydrochloride, N-(Ethyl-2,2,2-d3)ethan-2,2,2-d3-amine hydrochloride, Diethylamine-2,2,2,2',2',2'-D6 hydrochloride, Diethylamine-2,2,2,2',2',2'-D6 hydrochloride |
| InChI Formula | InChI=1 S/C4H11N.ClH/c1-3-5-4-2;/h5H,3-4H2,1-2H3;1 H/i1D3,2D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])CNCC([2 H])([2 H])[2 H] |
| IUPAC Name | 2,2,2-trideuterio-N-(2,2,2-trideuterioethyl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 115.6348 |
| Formula Weight | 115.1035 g/mol |
| Show More |