Learn More
Didodecylmethylamine, TRC
Specifications
Specifications
| Chemical Name or Material | Didodecylmethylamine |
| CAS | 2915-90-4 |
| Molecular Formula | C25 H53 N |
| Quantity | 100 g |
| Synonym | N-Dodecyl-N-methyl-1-dodecanamine, Dilaurylmethylamine, Dilaurylmonomethylamine, Farmin M 2-2095, Farmin M2 20, Methyldidodecylamine, N,N-Didodecyl-N-methylamine, N,N-Didodecylmethylamine, N-Dodecyl-N-methyldodecan-1-amine, N-Methyl-N,N-didodecylamine, N-Methyldidodecylamine, N-Dodecyl-N-methyldodecan-1-amine |
| InChI Formula | InChI=1S/C25H53N/c1-4-6-8-10-12-14-16-18-20-22-24-26(3)25-23-21-19-17-15-13-11-9-7-5-2/h4-25H2,1-3H3 |
| SMILES | CCCCCCCCCCCCN(C)CCCCCCCCCCCC |
| IUPAC Name | N-dodecyl-N-methyldodecan-1-amine |
| Molecular Weight (g/mol) | 367.695 |
| Formula Weight | 367.418 |