missing translation for 'onlineSavingsMsg'
Learn More

Didesethyl Chloroquine Hydroxyacetamide-d4, TRC

Product Code. 30373437
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373437 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30373437 Supplier Toronto Research Chemicals Supplier No. TRCD4409821MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Didesethyl Chloroquine Hydroxyacetamide 1,1,2,2-pentyl-d4
CAS 1216956-86-3
Molecular Formula C16 D4 H16 Cl N3 O2
Quantity 1 mg
Synonym 4-(4'-Glycolamidyl-1'-methyl-3,4-d4-butylamino)-7-chloroquinoline
InChI Formula InChI=1S/C16H20ClN3O2/c1-11(3-2-7-19-16(22)10-21)20-14-6-8-18-15-9-12(17)4-5-13(14)15/h4-6,8-9,11,21H,2-3,7,10H2,1H3,(H,18,20)(H,19,22)/i2D2,7D2
SMILES [2H]C([2H])(CC(C)Nc1ccnc2cc(Cl)ccc12)C([2H])([2H])NC(=O)CO
IUPAC Name N-[4-[(7-chloro-4-quinolyl)amino]-1,1,2,2-tetradeuterio-pentyl]-2-hydroxy-acetamide
Molecular Weight (g/mol) 325.83
Formula Weight 325.1495
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.