Learn More
Dicyclohexyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-cyclohexyl ester D4 |
| CAS | 358731-25-6 |
| Molecular Formula | C20 2H4 H22 O4 |
| Quantity | 0.05 g |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dicyclohexyl ester, Phthalic acid, bis-cyclohexyl ester D4, Dicyclohexyl phthalate-d4 |
| InChI Formula | InChI=1 S/C20H26O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h7-8,13-16 H,1-6,9-12H2/i7D,8 D,13 D,14 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OC2CCCCC2)c(C(=O)OC3CCCCC3)c1[2 H] |
| IUPAC Name | dicyclohexyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 334.44 |
| Formula Weight | 334.2082 g/mol |
| Show More |