Learn More
Dichloroacetic acid, 99%
Product Code. p-7022119
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
Product Code. 11447786
Brand: Thermo Scientific Alfa Aesar A14740.36
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 79-43-6 | C2H2Cl2O2 | 128.936 g/mol
Dichloroacetic acid salts are used as drugs due to its inhibition of the enzyme pyruvate dehydrogenase kinase. It is also employed to stem the production of lactic acid to treat lactic acidosis. It is clinically essential in children to treat lactic acidosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Miscible with water, ethanol, diethyl ether, n-octanol and acetone.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79-43-6
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.936
MDL Number MFCD00004223
InChI Key JXTHNDFMNIQAHM-UHFFFAOYSA-N
Synonym dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
PubChem CID 6597
ChEBI CHEBI:36386
IUPAC Name 2,2-dichloroacetic acid
SMILES C(C(=O)O)(Cl)Cl
Specifications
Melting Point 9°C to 11°C
Density 1.564
Boiling Point 192°C to 194°C
Flash Point >110°C (230°F)
Assay Percent Range 99%
Refractive Index 1.462
Quantity 500 g
UN Number UN1764
Beilstein 1098596
Sensitivity Hygroscopic
Merck Index 14,3050
Solubility Information Miscible with water,ethanol,diethyl ether,n-octanol and acetone.
Formula Weight 128.94
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dichloroacetic Acid