Learn More
Dichlofenthion, TRC
Specifications
Specifications
| Chemical Name or Material | Dichlofenthion |
| CAS | 97-17-6 |
| Molecular Formula | C10 H13 Cl2 O3 P S |
| Quantity | 50 mg |
| Synonym | Dichlofenthion, ECP, ECP (pesticide), ENT 17470, Hexanema, NSC 52964, Nemacide, O,O-Diethyl O-(2,4-dichlorophenyl) phosphorothioate, O-(2,4-Dichlorophenyl) O,O-diethyl phosphorothioate, VC 1-13, VC 13, VC 13 (pesticide), VC 13 Nemacide |
| InChI Formula | InChI=1S/C10H13Cl2O3PS/c1-3-13-16(17,14-4-2)15-10-6-5-8(11)7-9(10)12/h5-7H,3-4H2,1-2H3 |
| SMILES | CCOP(=S)(OCC)Oc1ccc(Cl)cc1Cl |
| IUPAC Name | (2,4-dichlorophenoxy)-diethoxy-sulfanylidene-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 315.15 |
| Formula Weight | 313.97 |