Learn More
Novus Biologicals™ Dicer knockout Mouse embryonic stem cells Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
0.25mg
Description
The Dicer knockout Mouse embryonic stem cells Lysate is derived from mouse. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| For Use With (Application) | Immunoprecipitation, Western Blot |
| Tissue | Embryo |
| Quantity | 0.25 mg |
| Storage Buffer | Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS. No reducing agents or loading buffer added. |
| Purity | Protein |
| Cellular Fraction | Whole |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |