Learn More
Olympus™ DIC Analyzer Cube, MDICT3
Click to view available options
For Use With (Equipment):
BX61, BX51, BX41 microscopes.
Unit Size:
Each
Description
- For differential interface contrast microscopy
- Filter cube mounting
Specifications
Specifications
| Type | DIC |
| For Use With (Equipment) | BX61, BX51, BX41 microscopes. |