Learn More
Olympus™ DIC Analyzer Cube, MDICT3
Product Code. 12621561
Click to view available options
For Use With (Equipment):
BX61, BX51, BX41 microscopes.
Unit Size:
Each
Product Code. 12621561
Brand: Olympus™ K10017446
Perform differential interface contrast microscopy using this analyzer. Olympus™ DIC Analyzer Cube, MDICT3 is conveniently mounted in a filter cube.
  • For differential interface contrast microscopy
  • Filter cube mounting
Specifications
Type DIC
For Use With (Equipment) BX61, BX51, BX41 microscopes.