Learn More
Dibromo Malonamide-13C3, TRC
Specifications
Specifications
| Chemical Name or Material | Dibromo Malonamide-13C3 |
| CAS | 1246815-05-3 |
| Molecular Formula | 13C3H4Br2N2O2 |
| Quantity | 10 mg |
| Synonym | 2,2-Dibromopropanediamide-13C3, 2,2-Dibromomalonamide-13C3, Dibromomalonic Acid Diamide-13C3 |
| InChI Formula | InChI=1S/C3H4Br2N2O2/c4-3(5,1(6)8)2(7)9/h(H2,6,8)(H2,7,9)/i1+1,2+1,3+1 |
| SMILES | N[13C](=O)[13C](Br)(Br)[13C](=O)N |
| IUPAC Name | 2,2-dibromo(1,2,3-^{13}C_{3})propanediamide |
| Molecular Weight (g/mol) | 262.86 |
| Formula Weight | 260.874 |