Learn More
Gilson™ DIAMOND™ Steril-Pack™ Sterile Pipet Tips
Traceable from the mould to the laboratory with batch number on each pack and rack. Diamond™ Steril-Pack™ Sterile Pipet Tips can be purchased sterilized or for autoclaving.
Brand:  Gilson™ F161330
93.64 EUR valid until 2024-12-31
Use promo code "23279" to get your promotional price.
Additional Details : Weight : 0.36000kg
 View more versions of this product
Product Code. 10307712
  • 102.00€ / Pack of 400
In Stock
Description
  • Tips comply with the CE IVD directive
  • Color coded packaging for volume recognition at a glance
  • Sterilized tips are free of detectable DNA, RNA, DNase, RNase, ATP, pyrogens and trace metals
  • Pre-sterilized tips are wrapped in medical device quality plastic
  • Gamma irradiated
  • Non-sterile tips can be autoclaved at 121°C for 20 minutes at 1 bar (moist heat)
Specifications
Specifications
Pipette tip
Sterile
D200
-0.075 to -0.05μL
5.1 cm
DNA, RNA, DNase, RNase, ATP, Pyrogen-free and trace Metals free
Universal
400 Pk.
Gilson Pipetman F20L, Gilson Pipetman F25L, Gilson Pipetman F50L, Gilson Pipetman F100L, Gilson Pipetman F200L, Gilson Pipetman P200L, Gilson Pipetman P20M, Gilson Pipetman P200M, Gilson Pipetman
Diamond Sterlipack
2 to 200 μL
Autoclavable
0.006 to 0.008μL
2 in.
CE, IVD
Liquid handling, medical, pharmaceutical and food industries, in various fields and applications such as PCR, molecular biology or cell technology
Box
Pipet Tips