Learn More
Scotch-Brite™ Diamond Floor Pads, Purple
Product Code. 17168790
Click to view available options
Diameter (Metric):
254 mm
280 mm
305 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
685 mm
Unit Size:
Pack of 5
Product Code. 17168790
Brand: Scotch-Brite™ 7100159499
Only null left
Add to basket
Add to basket
Diamond-coated floor pads for cleaning and polishing marble, terrazzo, slate, and polished concrete flooring. Helps eliminate floor finish and reduce labor costs.
  • Simultaneously cleans and polishes flooring to achieve an outstanding shine
  • Ideal for marble, terrazzo, vinyl, slate, and polished concrete floors
  • Easy to use: no expensive or harmful chemicals needed, only water
  • Recommended for use with low and standard speed machines
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Pads
Color Purple
Diameter (Metric) 432 mm
For Use With (Application) Burnishing, Cleaning
For Use With (Equipment) Marble, Terrazzo, Slate, and Polished Concrete Flooring
Material Diamond-coated
Style Round