missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diacetamide-d7, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | N-Acetylacetamide-D7 |
| CAS | 33945-50-5 |
| Molecular Formula | C4 D7 N O2 |
| Quantity | 0.1 g |
| Synonym | Acetamide-N,2,2,2-d4, N-(acetyl-d3)- (9 CI), Diacetamide-d7 (8 CI), N-Acetylacetamide-D7, N-(Acetyl-d3)acetamide-N,2,2,2-d4 |
| InChI Formula | InChI=1 S/C4H7NO2/c1-3(6)5-4(2)7/h1-2H3,(H,5,6,7)/i1D3,2D3/hD |
| SMILES | [2 H]N(C(=O)C([2 H])([2 H])[2 H])C(=O)C([2 H])([2 H])[2 H] |
| IUPAC Name | N,2,2,2-tetradeuterio-N-(2,2,2-trideuterioacetyl)acetamide |
| Molecular Weight (g/mol) | 108.147 |
| Formula Weight | 108.0916 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction