Learn More
Di-tert-butyl Ether, TRC
Specifications
Specifications
| Chemical Name or Material | Di-tert-butyl Ether |
| CAS | 6163-66-2 |
| Molecular Formula | C8H18O |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C8H18O/c1-7(2,3)9-8(4,5)6/h1-6H3 |
| SMILES | CC(C)(C)OC(C)(C)C |
| IUPAC Name | 2-methyl-2-[(2-methylpropan-2-yl)oxy]propane |
| Molecular Weight (g/mol) | 130.23 |
| Formula Weight | 130.1358 |