Learn More
Di-n-octyl Phthalate, TRC
Product Code. 17914995
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17914995 1 g 1g
30004567 50 g 50g
30007171 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17914995 Supplier Toronto Research Chemicals Supplier No. TRCD4817501G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 117-84-0
Molecular Formula C24 H38 O4
Molecular Weight (g/mol) 390.56
Synonym 1,2-Benzenedicarboxylic acid, 1,2-dioctyl ester, 1,2-Benzenedicarboxylic acid, dioctyl ester (9CI), Phthalic acid, dioctyl ester (6CI,8CI), 1,2-Di-n-octyl phthalate, Bis(n-octyl) phthalate, DNOP, Di-n-octyl phthalate, Dicaprylyl phthalate, Dinopol NOP, Dioctyl o-phthalate, Dioctyl phthalate, Monocizer W 555, NSC 15318, Octyl phthalate, Phthalic acid dioctyl ester, Vinycizer 85, n-Octyl phthalate, Phthalic acid di-n-octyl ester, Phthalic acid, bis-n-octyl ester
IUPAC Name dioctyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC
Specifications
Quantity 1 g
InChI Formula InChI=1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3
Formula Weight 390.277
Chemical Name or Material Di-n-octyl phthalate