missing translation for 'onlineSavingsMsg'
Learn More

Di-D-fructofuranose 1,2':2,1'-Dianhydride, TRC

Product Code. 30376943
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376943 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376943 Supplier Toronto Research Chemicals Supplier No. TRCD443960100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-D-fructofuranose 1,2':2,1'-dianhydride
CAS 3568-49-8
Molecular Formula C12 H20 O10
Quantity 100 mg
InChI Formula InChI=1S/C12H20O10/c13-1-5-7(15)9(17)11(21-5)3-20-12(4-19-11)10(18)8(16)6(2-14)22-12/h5-10,13-18H,1-4H2/t5-,6-,7-,8-,9+,10+,11?,12?/m1/s1
SMILES O[C@@H]1C2(O[C@H](CO)[C@H]1O)COC3(CO2)O[C@H](CO)[C@@H](O)[C@@H]3O
IUPAC Name (1S,2S,3R,9S,10S,11R)-3,11-bis(hydroxymethyl)-4,7,12,14-tetraoxadispiro[4.2.48.25]tetradecane-1,2,9,10-tetrol
Molecular Weight (g/mol) 324.281
Formula Weight 324.106
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.