Learn More
Di(Alpha-tocopherol) Phosphate, TRC
Product Code. 30452852
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452852 100 mg 100mg
30452213 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30452852 Supplier Toronto Research Chemicals Supplier No. TRCT526190100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 311313-37-8
Molecular Formula C58H99O6P
Molecular Weight (g/mol) 923.38
Synonym (2R,2'R)-3,4-Dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-ol 6,6'-(Hydrogen Phosphate)
IUPAC Name bis[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] hydrogen phosphate
SMILES CC1=C(C)C(OP(OC2=C(C)C(CC[C@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)O3)=C3C(C)=C2C)(O)=O)=C(C)C4=C1O[C@@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)CC4
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C58H99O6P/c1-39(2)23-17-25-41(5)27-19-29-43(7)31-21-35-57(15)37-33-51-49(13)53(45(9)47(11)55(51)61-57)63-65(59,60)64-54-46(10)48(12)56-52(50(54)14)34-38-58(16,62-56)36-22-32-44(8)30-20-28-42(6)26-18-24-40(3)4/h39-44H,17-38H2,1-16H3,(H,59,60)/t41-,42-,43-,44-,57-,58-/m1/s1
Formula Weight 922.7179
Chemical Name or Material Di(alpha-tocopherol) Phosphate