Learn More
Di-4-ANBDQPQ (Mixture of E/Z isomers, ~90%), TRC
Product Code. 17999724
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
10 mg
Unit Size:
0.5mg
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17999724 0.5 mg 0.5mg
17979064 10 mg 10mg
17988204 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17999724 Supplier Toronto Research Chemicals Supplier No. TRCD4164330.5MG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1005388-96-4
Molecular Formula C37H49N3 • 2(Br)
Molecular Weight (g/mol) 535.802799
Synonym 4-[4-[6-(Dibutylamino)-2-naphthalenyl]-1,3-butadien-1-yl]-1-[3-(trimethylammonio)propyl]quinolinium Bromide, JPW 6003
SMILES C[N+](C)(CCC[N+]1=CC=C(/C=C/C=C\C2=CC3=C(C=C(C=C3)N(CCCC)CCCC)C=C2)C4=C1C=CC=C4)C.[Br-].[Br-]
Specifications
Quantity 0.5 mg
InChI Formula InChI=1S/C37H49N3.2BrH/c1-6-8-24-38(25-9-7-2)35-22-21-33-29-31(19-20-34(33)30-35)15-10-11-16-32-23-27-39(26-14-28-40(3,4)5)37-18-13-12-17-36(32)37;;/h10-13,15-23,27,29-30H,6-9,14,24-26,28H2,1-5H3;2*1H/q+2;;/p-2
Chemical Name or Material Di-4-ANBDQPQ (Mixture of E/Z isomers)