missing translation for 'onlineSavingsMsg'
Learn More

Dextrinized Corn Starch, MP Biomedicals

Product Code. 11451765 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
5 lb.
25 lb.
Unit Size:
25lb
5lb
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451765 5 lb. 5lb
11461765 25 lb. 25lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451765 Supplier MP Biomedicals Supplier No. 0296042905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • A component in animal diet

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym Maltodextin

Specifications

Color White
Quantity 5 lb.
Physical Form Powder
Chemical Name or Material DEXTRINIZED CORN STARCH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.