Learn More
Dextran, Pure, 150, M.W. 100,000-200,000, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
:
100g, Plastic powder jar
Specifications
Specifications
| Color | White |
| CAS | 9004-54-0 |
| Physical Form | Solid |
| MDL Number | 130935 |
| Quantity | 100 g |
| Packaging | Plastic powder jar |